Invitrogen
Manufacturer Code:PA536153
Catalog # PIPA536153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 500-550 of human UBF-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 90-kDa nucleolus organizer region autoantigen; Autoantigen NOR-90; Nucleolar transcription factor 1; Nucleolar transcription factor 1 Autoantigen NOR-90 Upstream-binding factor 1 UBF-1 UBTF UBF UBF1; transcription factor UBF; UBF-1; Upstream-binding factor 1
Gene Aliases: A930005G04Rik; NOR-90; Tcfubf; UBF; UBF-1; UBF1; UBF2; UBTF
UniProt ID: (Human) P17480, (Rat) P25977
Entrez Gene ID: (Human) 7343, (Rat) 25574, (Mouse) 21429
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.