Invitrogen
Manufacturer Code:PA555552
Catalog # PIPA555552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UBE3D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase E3D; HECT-type E3 ubiquitin transferase E3D; UBCH10 binding protein with a hect-like domain; UbcH10-binding protein with a HECT-like domain; ubiquitin-conjugating enzyme E2C binding protein; Ubiquitin-conjugating enzyme E2C-binding protein
Gene Aliases: C6orf157; H10BH; UBE2CBP; UBE3D; YJR141W
UniProt ID: (Human) Q6IPR4
Entrez Gene ID: (Human) 90025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.