Abnova
Manufacturer Code:PAB21883
Catalog # 89-123-587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human UBE3D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant UBE3D.
For Research Use Only
Protein Aliases: C6orf157; E3 ubiquitin-protein ligase E3D; H10BH; HECT-type E3 ubiquitin transferase E3D; RP4-751H9.1; UBCH10 binding protein with a hect-like domain; UbcH10-binding protein with a HECT-like domain; UBE2CBP; ubiquitin-conjugating enzyme E2C binding protein; Ubiquitin-conjugating enzyme E2C-binding protein; YJR141W
Gene Aliases: C6orf157; H10BH; UBE2CBP; UBE3D; YJR141W
UniProt ID: (Human) Q6IPR4
Entrez Gene ID: (Human) 90025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.