Invitrogen
Manufacturer Code:PA535265
Catalog # PIPA535265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 549-578 amino acids from the central region of human UBE3C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HECT-type ubiquitin transferase E3C; HectH2; UBE3CKIAA0010 KIAA10 Ubiquitin-protein ligase E3C Ubiquitin-protein ligase E3C HectH2; ubiquitin-protein isopeptide ligase (E3); Ubiquitin-protein ligase E3C
Gene Aliases: HECTH2; KIAA0010; KIAA10; UBE3C
UniProt ID: (Human) Q15386
Entrez Gene ID: (Human) 9690
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.