Novus Biologicals
Manufacturer Code:NBP29402320UL
Catalog # NB022611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human UBE3C (NP_055486.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.3.2.- KIAA0010KIAA10ubiquitin-protein isopeptide ligase (E3) ubiquitin protein ligase E3C ubiquitin-protein ligase E3C; HECT-type ubiquitin transferase E3C; HectH2; ubiquitin-protein isopeptide ligase (E3); Ubiquitin-protein ligase E3C
Gene Aliases: HECTH2; KIAA0010; KIAA10; UBE3C
UniProt ID: (Human) Q15386
Entrez Gene ID: (Human) 9690
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.