Proteintech
Manufacturer Code:123331AP150UL
Catalog # 50-172-7902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBE3C Fusion Protein Ag2966 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HECT-type ubiquitin transferase E3C; HectH2; KIAA0010 KIAA10 UBE3C ubiquitin protein ligase E3C; ubiquitin protein ligase E3C; ubiquitin-protein isopeptide ligase (E3); Ubiquitin-protein ligase E3C
Gene Aliases: AI853514; HECTH2; KIAA0010; KIAA10; mKIAA0010; RGD1559986; UBE3C
UniProt ID: (Human) Q15386, (Mouse) Q80U95
Entrez Gene ID: (Human) 9690, (Rat) 362294, (Mouse) 100763
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.