Proteintech
Manufacturer Code:103441AP150UL
Catalog # 50-172-6230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBE3A Fusion Protein Ag0346 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANCR AS E6 AP E6AP E6AP ubiquitin protein ligase EPVE6AP HPVE6A UBE3A ubiquitin protein ligase E3A; CTCL tumor antigen se37-2; E6-AP ubiquitin protein ligase; E6AP ubiquitin-protein ligase; HECT-type ubiquitin transferase E3A; human papilloma virus E6-associated protein; Human papillomavirus E6-associated protein; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; ubiquitin conjugating enzyme E3A; Ubiquitin-protein ligase E3A
Gene Aliases: 4732496B02; 5830462N02Rik; A130086L21Rik; ANCR; AS; E6-AP; E6AP; EPVE6AP; HPVE6A; mKIAA4216; UBE3A
UniProt ID: (Human) Q05086, (Mouse) O08759
Entrez Gene ID: (Human) 7337, (Rat) 361585, (Mouse) 22215
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.