Invitrogen
Manufacturer Code:PA141467
Catalog # 01-675-295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 12-28 of human UBE2V2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1 alpha,25-dihydroxyvitamin D3-inducible; DDVit 1; DDVIT1 DDVit-1 EDAF-1 EDPF-1 EDPF1 MMS2 UEV-2 UEV2; EDAF-1; EDPF-1; enterocyte differentiation promoting factor; Enterocyte differentiation-associated factor 1; enterocyte differentiation-associated factor EDAF-1; Enterocyte differentiation-promoting factor 1; methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of; MMS2 homolog; ubiquitin conjugating enzyme E2 variant 2; Ubiquitin-conjugating enzyme E2 variant 2; Vitamin D3-inducible protein
Gene Aliases: DDVit-1; DDVIT1; EDAF-1; EDPF-1; EDPF1; MMS2; UBE2V2; UEV-2; UEV2
UniProt ID: (Human) Q15819
Entrez Gene ID: (Human) 7336
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.