Novus Biologicals
Manufacturer Code:NBP274750JF646
Catalog # N274750J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F5 |
Immunogen | Full length recombinant protein of human UBE2T (NP_054895) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cell proliferation-inducing gene 50 protein; Cell proliferation-inducing gene 50 protein EC 6.3.2.19 HSPC150 HSPC150 protein similar to ubiquitin-conjugating enzyme PIG50 Ubiquitin carrier protein T ubiquitin conjugating enzyme ubiquitin-conjugating enzyme E2 T ubiquitin-conjugating enzyme E2T (putative) Ubiquitin-protein ligase T; E2 ubiquitin-conjugating enzyme T; HSPC150 protein similar to ubiquitin-conjugating enzyme; Ubiquitin carrier protein T; ubiquitin conjugating enzyme E2T; Ubiquitin-conjugating enzyme E2 T; ubiquitin-conjugating enzyme E2T (putative); Ubiquitin-protein ligase T
Gene Aliases: FANCT; HSPC150; PIG50; UBE2T
UniProt ID: (Human) Q9NPD8
Entrez Gene ID: (Human) 29089
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.