Invitrogen
Manufacturer Code:PA576202
Catalog # PIPA576202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human UBE2T. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 50 protein; E2 ubiquitin-conjugating enzyme T; FANCT PIG50 HSPC150; HSPC150 protein similar to ubiquitin-conjugating enzyme; Ubiquitin carrier protein T; ubiquitin conjugating enzyme E2T; Ubiquitin-conjugating enzyme E2 T; ubiquitin-conjugating enzyme E2T (putative); Ubiquitin-protein ligase T
Gene Aliases: 2700084L22Rik; C80607; FANCT; HSPC150; PIG50; RGD1310816; UBE2T
UniProt ID: (Human) Q9NPD8, (Mouse) Q9CQ37
Entrez Gene ID: (Human) 29089, (Rat) 360847, (Mouse) 67196
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.