Invitrogen
Manufacturer Code:PA570600
Catalog # PIPA570600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human UBE2Q1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme Q1; Galactosyl transferase-associated protein; GTAP; Protein NICE-5; Ubiquitin carrier protein Q1; ubiquitin conjugating enzyme E2Q family member 1; Ubiquitin-conjugating enzyme E2 Q1; ubiquitin-conjugating enzyme E2Q (putative) 1; ubiquitin-conjugating enzyme E2Q family member 1; Ubiquitin-protein ligase Q1
Gene Aliases: 1110002C01Rik; 2310012M18Rik; AI854014; GTAP; NICE-5; NICE5; PRO3094; UBE2Q; UBE2Q1
UniProt ID: (Human) Q7Z7E8, (Mouse) Q7TSS2
Entrez Gene ID: (Pig) 100156747, (Dog) 100688676, (Horse) 100062653, (Rabbit) 100347500, (Human) 55585, (Bovine) 505133, (Mouse) 70093, (Rat) 295252, (Guinea pig) 100732687
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.