Abnova
Manufacturer Code:H00055585K
Catalog # 89-210-614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human UBE2Q1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human UBE2Q1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme Q1; galactosyl transferase-associated protein; GTAP NICE-5 PRO3094 UBE2Q; Protein NICE-5; Ubiquitin carrier protein Q1; ubiquitin conjugating enzyme E2Q family member 1; Ubiquitin-conjugating enzyme E2 Q1; ubiquitin-conjugating enzyme E2Q (putative) 1; ubiquitin-conjugating enzyme E2Q family member 1; Ubiquitin-protein ligase Q1
Gene Aliases: GTAP; NICE-5; NICE5; PRO3094; UBE2Q; UBE2Q1
UniProt ID: (Human) Q7Z7E8
Entrez Gene ID: (Human) 55585
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.