Invitrogen
Manufacturer Code:PA576376
Catalog # PIPA576376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human UBE2NL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epididymis tissue protein Li 174; Li174 Putative ubiquitin-conjugating enzyme E2 N-like; Putative ubiquitin-conjugating enzyme E2 N-like; ubiquitin conjugating enzyme E2N-like; ubiquitin-conjugating enzyme E2N-like
Gene Aliases: Li174; UBE2NL
UniProt ID: (Human) Q5JXB2
Entrez Gene ID: (Human) 389898
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.