Novus Biologicals
Manufacturer Code:NBP176593SS
Catalog # NBP176593SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to 15 amino acids near the C-terminus of human UBC13. The immunogen is located within the last 50 amino acids of UBC13. Amino Acid Squence: NPDDPLANDVAEQWK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: bendless-like ubiquitin conjugating enzyme; bendless-like ubiquitin conjugating enzyme Bendless-like ubiquitin-conjugating enzyme BLU EC 6.3.2.19 MGC131857 MGC8489 UBC13 UbcH-ben Ubiquitin carrier protein N ubiquitin-conjugating enzyme E2 N ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13) ubiquitin-conjugating enzyme E2N (UBC13 homolog yeast) Ubiquitin-protein ligase N yeast UBC13 homolog; Bendless-like ubiquitin-conjugating enzyme; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; Ubc13; UbcH13; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2N; Ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); Ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Aliases: BLU; HEL-S-71; UBC13; UbcH-ben; UbcH13; UBCHBEN; UBC13; UBE2N
UniProt ID: (Human) P61088
Entrez Gene ID: (Human) 7334
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.