Invitrogen
Manufacturer Code:PA511969
Catalog # PIPA511969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 123-153 amino acids from the C-terminal region of human UBE2L6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme L6; retinoic acid induced gene B protein; Retinoic acid-induced gene B protein; RIG-B; UbcH8; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2L 6; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6; Ubiquitin/ISG15-conjugating enzyme E2 L6; Ubiquitin/ISG15-conjugating enzyme E2 L6 Ubiquitin-protein ligase L6 Ubiquitin carrier protein L6 UbcH8 Retinoic acid-induced gene B protein RIG-B
Gene Aliases: RIG-B; UBCH8; UBE2L6
UniProt ID: (Human) O14933
Entrez Gene ID: (Human) 9246
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.