Abnova
Manufacturer Code:PAB29396
Catalog # 89-935-074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human UBE2L6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant human UBE2L6.
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme L6; MGC40331; retinoic acid induced gene B protein; Retinoic acid-induced gene B protein; RIG-B; UBCH8; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2L 6; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6; Ubiquitin/ISG15-conjugating enzyme E2 L6
Gene Aliases: RIG-B; UBCH8; UBE2L6
UniProt ID: (Human) O14933
Entrez Gene ID: (Human) 9246
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.