Invitrogen
Manufacturer Code:PA592154
Catalog # PIPA592154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 73-212 of human UBE2L3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme L3; L-UBC; UbcH7; UbcM4; Ubiquitin carrier protein L3; ubiquitin conjugating enzyme E2L 3; ubiquitin-conjugating enzyme 7; Ubiquitin-conjugating enzyme E2 L3; Ubiquitin-conjugating enzyme E2-F1; ubiquitin-conjugating enzyme E2L 3; ubiquitin-conjugating enzyme UBCH7; Ubiquitin-protein ligase L3
Gene Aliases: C79827; E2-F1; L-UBC; UBCE7; UBCH7; UbcM4; UBE2L3
UniProt ID: (Human) P68036, (Mouse) P68037
Entrez Gene ID: (Human) 7332, (Rat) 363836, (Mouse) 22195
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.