Invitrogen
Manufacturer Code:PA577201
Catalog # PIPA577201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human UBE2J2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme J2; NCUBE-2; Non-canonical ubiquitin-conjugating enzyme 2; Ubc6p homolog; ubiquitin conjugating enzyme 6; ubiquitin conjugating enzyme E2, J2; Ubiquitin-conjugating enzyme E2 J2; Ubiquitin-conjugating enzyme E2 J2 Non-canonical ubiquitin-conjugating enzyme 2 NCUBE-2 UBE2J2 NCUBE2; ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J2 homolog; yeast UBC6 homolog
Gene Aliases: 1200007B18Rik; 2400008G19Rik; 5730472G04Rik; AL022923; NCUBE-2; NCUBE2; PRO2121; Ubc6; Ubc6p; UBE2J2
UniProt ID: (Human) Q8N2K1, (Mouse) Q6P073
Entrez Gene ID: (Human) 118424, (Mouse) 140499, (Rat) 298689
Molecular Function: transfer/carrier protein ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.