Abnova
Manufacturer Code:PAB16908
Catalog # 89-118-720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human UBE2J2 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of UBE2J2 .
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme J2; NCUBE-2; NCUBE2; Non-canonical ubiquitin-conjugating enzyme 2; PRO2121; ubiquitin conjugating enzyme 6; ubiquitin conjugating enzyme E2, J2; Ubiquitin-conjugating enzyme E2 J2; ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast); yeast UBC6 homolog
Gene Aliases: NCUBE-2; NCUBE2; PRO2121; UBE2J2
UniProt ID: (Human) Q8N2K1
Entrez Gene ID: (Human) 118424
Molecular Function:
transfer/carrier protein
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.