Novus Biologicals
Manufacturer Code:NBP300096B
Catalog # NB086703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant UBE2I/Ubc9 (P63279 Met1-Ser158). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C358B7.1 EC 6.3.2 EC 6.3.2.- P18 SUMO-1-protein ligase SUMO-protein ligase UBC9SUMO-conjugating enzyme UBC9 UBCE9 Ubiquitin carrier protein 9 Ubiquitin carrier protein I ubiquitin conjugating enzyme 9 Ubiquitin-conjugating enzyme E2 I ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9) ubiquitin-conjugating enzyme E2I (UBC9 homolog yeast) ubiquitin-conjugating enzyme UbcE2A ubiquitin-like protein SUMO-1 conjugating enzyme ubiquitin-protein ligase E2I Ubiquitin-protein ligase I; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; Ubiquitin carrier protein 9; Ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2I; Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; Ubiquitin-protein ligase I
Gene Aliases: C358B7.1; P18; UBC9; UBCE9; UBE2I
UniProt ID: (Human) P63279
Entrez Gene ID: (Human) 7329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.