Proteintech
Manufacturer Code:170561AP150UL
Catalog # 50-173-1354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBE2F Fusion Protein Ag10764 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NCE2 NEDD8 carrier protein UBE2F NEDD8 conjugating enzyme 2 NEDD8 protein ligase UBE2F UBE2F; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RING-type E3 NEDD8 transferase UBE2F; ubiquitin conjugating enzyme E2F (putative); Ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative)
Gene Aliases: 2510010F15Rik; AI851109; NCE2; RGD1307608; UBE2F; Ube2fa
UniProt ID: (Human) Q969M7, (Rat) Q5U203, (Mouse) Q9CY34
Entrez Gene ID: (Human) 140739, (Rat) 363284, (Mouse) 67921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.