OriGene
Manufacturer Code:CF800072
Catalog # 50-166-4835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6D10 |
Immunogen | Full length human recombit protein of human UBE2E3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme E3; UbcH9; UBCH9 UbcM2; UbcM2; Ubiquitin carrier protein E3; ubiquitin conjugating enzyme E2E 3; ubiquitin-conjugating enzyme 4; Ubiquitin-conjugating enzyme E2 E3; Ubiquitin-conjugating enzyme E2-23 kDa; ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 3, UBC4/5 homolog; Ubiquitin-protein ligase E3
Gene Aliases: UBCE4; UBCH9; UbcM2; UBE2E3
UniProt ID: (Human) Q969T4, (Mouse) P52483
Entrez Gene ID: (Human) 10477, (Rat) 295686, (Mouse) 22193
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.