Proteintech
Manufacturer Code:118441AP150UL
Catalog # 50-172-7501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBE2E2 Fusion Protein Ag2509 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme E2; TBC1 domain family, member 12; UbcH8; UBCH8 UBE2E2 Ubiquitin carrier protein E2 Ubiquitin protein ligase E2; Ubiquitin carrier protein E2; ubiquitin conjugating enzyme E2E 2; Ubiquitin-conjugating enzyme E2 E2; ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast); Ubiquitin-protein ligase E2
Gene Aliases: BC016265; UBCH8; UBE2E2
UniProt ID: (Human) Q96LR5, (Mouse) Q91W82
Entrez Gene ID: (Human) 7325, (Rat) 361013, (Mouse) 218793
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.