Invitrogen
Manufacturer Code:PA542280
Catalog # PIPA542280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human UBE2D3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme D3; E2 ubiquitin-conjugating enzyme D3; E2(17)KB 3; PAPase; Phosphoarginine phosphatase; PRO2116; Ubiquitin carrier protein D3; ubiquitin conjugating enzyme E2D 3; Ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2(17)KB 3; Ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); Ubiquitin-conjugating enzyme E2D 3 E2(17)KB3 MGC43926 MGC5416 UBC4/5 UBCH5C; Ubiquitin-protein ligase D3
Gene Aliases: 1100001F19Rik; 9430029A22Rik; AA414951; E2(17)KB3; eff; fb36f02; UBC4/5; UBC5C; UBCH5C; UBE2D3; UBE2D3P; wu:fb36f02; zgc:56340
UniProt ID: (Human) P61077, (Bovine) Q3ZCF7, (Mouse) P61079, (Rat) P61078
Entrez Gene ID: (Rabbit) 100347511, (Dog) 478495, (Horse) 100630481, (Human) 7323, (Bovine) 326583, (Mouse) 66105, (Rat) 81920, (Guinea pig) 100730907, (Zebrafish) 321832
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.