Invitrogen
Manufacturer Code:MA537771
Catalog # PIMA537771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2001CT211.37.61 |
Immunogen | KLH conjugated synthetic peptide between 137~172 amino acids from the C-terminal region of human UBE2C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme C; (E3-independent) E2 ubiquitin-conjugating enzyme C 1110015A16Rik cyclin-selective ubiquitin carrier protein D2Ertd695e dJ447F3.2 E2 ubiquitin-conjugating enzyme C mitotic-specific ubiquitin-conjugating enzyme RP3-447F3.1 ubcH10 UBE2C ubiquitin carrier protein C ubiquitin conjugating enzyme E2 C ubiquitin conjugating enzyme E2C ubiquitin-conjugating enzyme E2 C ubiquitin-conjugating enzyme E2C ubiquitin-protein ligase C; cyclin-selective ubiquitin carrier protein; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; UbcH10; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2C; Ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; Ubiquitin-protein ligase C
Gene Aliases: dJ447F3.2; UBCH10; UBE2C
UniProt ID: (Human) O00762
Entrez Gene ID: (Human) 11065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.