Invitrogen
Manufacturer Code:PA5111407
Catalog # PIPA5111407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human UBE2C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme C; (E3-independent) E2 ubiquitin-conjugating enzyme C 1110015A16Rik D2Ertd695e dJ447F3.2 E2 ubiquitin-conjugating enzyme C RP3-447F3.1 Ubch10 Ube2c Ubiquitin carrier protein C Ubiquitin-conjugating enzyme E2 C Ubiquitin-protein ligase C; cyclin-selective ubiquitin carrier protein; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; UbcH10; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2C; Ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; Ubiquitin-protein ligase C
Gene Aliases: dJ447F3.2; UBCH10; UBE2C
UniProt ID: (Human) O00762
Entrez Gene ID: (Human) 11065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.