Invitrogen
Manufacturer Code:PA521316
Catalog # PIPA521316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 182 of UBE2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme B; E2-17kDa HHR6B HR6B RAD6B UBC2 E2 protein RAD6 homolog B ubiquitin carrier protein B ubiquitin-conjugating enzyme E2 B ubiquitin-conjugating enzyme E2-17 kDa ubiquitin-conjugating enzyme E2B (RAD6 homolog) ubiquitin-protein ligase B; E214K; HR6B; RAD6 homolog B; Ubiquitin carrier protein B; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B, RAD6 homology; Ubiquitin-protein ligase B
Gene Aliases: 2610301N02Rik; E2-14k; E2-17kDa; HHR6B; HR6B; mHR6B; RAD6B; UBC2; UBE2B
UniProt ID: (Human) P63146, (Mouse) P63147
Entrez Gene ID: (Human) 7320, (Mouse) 22210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.