Abnova
Manufacturer Code:H00007320A01
Catalog # 89-105-694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 63 a.a. &bsim 152 a.a) partial recombinant protein with GST tag.,UBE2B (NP_003328.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant UBE2B.
For Research Use Only
Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme B; E2-17kDa; HHR6B; HR6B; RAD6 homolog B; RAD6B; UBC2; Ubiquitin carrier protein B; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B (RAD6 homolog); Ubiquitin-protein ligase B
Gene Aliases: E2-17kDa; HHR6B; HR6B; RAD6B; UBC2; UBE2B
UniProt ID: (Human) P63146
Entrez Gene ID: (Human) 7320
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.