Invitrogen
Manufacturer Code:PA5104442
Catalog # PIPA5104442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human UBE2A/B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610301N02Rik E2 protein E2 ubiquitin-conjugating enzyme A E2 ubiquitin-conjugating enzyme B E2(14k) E214K E2-14k E2-17kDa HHR6A HHR6B HR6A HR6B Mhr6a mHR6B mp:zf637-2-000771 MRXS30 MRXSN RAD6 homolog A RAD6 homolog B Rad6a Rad6b si:bz46j2.4 UBC2 UBE2A ube2b Ubiquitin carrier protein A Ubiquitin carrier protein B ubiquitin conjugating enzyme ubiquitin conjugating enzyme E2 A ubiquitin conjugating enzyme E2 B ubiquitin conjugating enzyme E2A ubiquitin conjugating enzyme E2B ubiquitin-conjugating enzyme E2 A Ubiquitin-conjugating enzyme E2 B ubiquitin-conjugating enzyme E2(14k) Ubiquitin-conjugating enzyme E2-17 kDa ubiquitin-conjugating enzyme E2A ubiquitin-conjugating enzyme E2A (RAD6 homolog) ubiquitin-conjugating enzyme E2A RAD6 homolog ubiquitin-conjugating enzyme E2B ubiquitin-conjugating enzyme E2B (RAD6 homolog) ubiquitin-conjugating enzyme E2B (RAD6 homolog S. cerevisiae) ubiquitin-conjugating enzyme E2B RAD6 homolog ubiquitin-conjugating enzyme E2B RAD6 homology ubiquitin-conjugating enzyme HR6A ubiquitin-protein ligase A Ubiquitin-protein ligase B wu:fa01h11 zgc:100921; E2 protein; E2 ubiquitin-conjugating enzyme A; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; HR6A; HR6B; RAD6 homolog A; RAD6 homolog B; Ubiquitin carrier protein A; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2A; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 A; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-conjugating enzyme HR6A; Ubiquitin-protein ligase A; Ubiquitin-protein ligase B
Gene Aliases: 2610301N02Rik; E2-14k; E2-17kDa; HHR6A; HHR6B; HR6A; HR6B; Mhr6a; mHR6B; MRXS30; MRXSN; RAD6A; RAD6B; UBC2; UBE2A; UBE2B
UniProt ID: (Human) P49459, (Human) P63146, (Mouse) Q9Z255, (Rat) P63149, (Mouse) P63147
Entrez Gene ID: (Human) 7319, (Human) 7320, (Rat) 298317, (Mouse) 22209, (Rat) 81816, (Mouse) 22210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.