Invitrogen
Manufacturer Code:PA5112140
Catalog # PIPA5112140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Rad6/UBE2A Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme A; E2 ubiquitin-conjugating enzyme A HHR6A HR6A Mhr6a mp:zf637-2-000771 MRXS30 MRXSN RAD6 homolog A Rad6a si:bz46j2.4 UBC2 Ube2a Ubiquitin carrier protein A ubiquitin conjugating enzyme E2 A ubiquitin conjugating enzyme E2A Ubiquitin-conjugating enzyme E2 A ubiquitin-conjugating enzyme E2A ubiquitin-conjugating enzyme E2A (RAD6 homolog) ubiquitin-conjugating enzyme E2A RAD6 homolog ubiquitin-conjugating enzyme HR6A Ubiquitin-protein ligase A wu:fa01h11; HR6A; RAD6 homolog A; Ubiquitin carrier protein A; ubiquitin conjugating enzyme E2A; Ubiquitin-conjugating enzyme E2 A; Ubiquitin-protein ligase A
Gene Aliases: HHR6A; MRXS30; MRXSN; RAD6A; UBC2; UBE2A
UniProt ID: (Human) P49459
Entrez Gene ID: (Human) 7319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.