Proteintech
Manufacturer Code:159121AP150UL
Catalog # 50-173-0569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBE1 Fusion Protein Ag8703 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1S9 A1S9T A1ST AMCX1 GXP1 Protein A1S9 SMAX2 UBA1 UBA1A UBE1 UBE1X Ubiquitin activating enzyme E1; A1S9T and BN75 temperature sensitivity complementing; POC20 centriolar protein homolog; Protein A1S9; testicular secretory protein Li 63; uba1 {ECO:0000312|EMBL:AAH85791.1, ECO:0000312|RGD:1359327}; UBA1, ubiquitin-activating enzyme E1 homolog A; Ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1 {ECO:0000250|UniProtKB:Q02053}; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1 X; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250|UniProtKB:Q02053}
Gene Aliases: A1S9; A1S9T; A1ST; AMCX1; CFAP124; GXP1; POC20; Sbx; SMAX2; UBA1; UBA1A; Ube-1; UBE1; Ube1ax; UBE1X
UniProt ID: (Human) P22314, (Mouse) Q02053, (Rat) Q5U300
Entrez Gene ID: (Human) 7317, (Mouse) 22201, (Rat) 314432
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.