Invitrogen
Manufacturer Code:MA515916
Catalog # PIMA515916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B10 |
Immunogen | Purified recombit fragment of human UBE2I expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C358B7.1 P18 UBC9 SUMO-1-protein ligase SUMO-conjugating enzyme UBC9 SUMO-protein ligase ubiquitin carrier protein 9 ubiquitin carrier protein I ubiquitin conjugating enzyme 9 ubiquitin-conjugating enzyme E2I (UBC9 homolog yeast) ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9) ubiquitin-conjugating enzyme UbcE2A ubiquitin-like protein SUMO-1 conjugating enzyme ubiquitin-protein ligase E2I ubiquitin-protein ligase I; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; Ubiquitin carrier protein 9; Ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2I; Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; Ubiquitin-protein ligase I
Gene Aliases: C358B7.1; P18; UBC9; UBCE9; UBE2I
UniProt ID: (Human) P63279
Entrez Gene ID: (Human) 7329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.