Invitrogen
Manufacturer Code:PA531003
Catalog # PIPA531003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 194 of Human UBC3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC34B E2-CDC34B UBC3B ubiquitin carrier protein R2 ubiquitin-conjugating enzyme E2 R2 ubiquitin-conjugating enzyme E2-CDC34B ubiquitin-conjugating enzyme UBC3B ubiquitin-protein ligase R2; E2 ubiquitin-conjugating enzyme R2; E2-CDC34B; Ubiquitin carrier protein R2; ubiquitin conjugating enzyme E2R 2; Ubiquitin-conjugating enzyme E2 R2; Ubiquitin-conjugating enzyme E2-CDC34B; ubiquitin-conjugating enzyme E2R 2; ubiquitin-conjugating enzyme UBC3B; Ubiquitin-protein ligase R2
Gene Aliases: 1200003M11Rik; CDC34B; E2-CDC34B; UBC3B; UBE2R2
UniProt ID: (Human) Q712K3, (Mouse) Q6ZWZ2
Entrez Gene ID: (Human) 54926, (Mouse) 67615
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.