Invitrogen
Manufacturer Code:MA534786
Catalog # PIMA534786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG38-68 |
Immunogen | Recombinant protein within human Ube2N aa 1-150. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1500026J17Rik AL022654 BB101821 Bendless-like ubiquitin-conjugating enzyme Blu E2 ubiquitin-conjugating enzyme N HEL-S-71 MGC131857 MGC8489 Ubc13 UbcH13 UbcH-ben UBCHBEN UBC13 UBE2N Ubiquitin carrier protein N Ubiquitin-conjugating enzyme E2 N Ubiquitin-protein ligase N; bendless-like ubiquitin conjugating enzyme; Bendless-like ubiquitin-conjugating enzyme; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; Ubc13; UbcH13; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2N; Ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); Ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Aliases: BLU; HEL-S-71; UBC13; UbcH-ben; UbcH13; UBCHBEN; UBC13; UBE2N
UniProt ID: (Human) P61088
Entrez Gene ID: (Human) 7334
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.