Proteintech
Manufacturer Code:102431AP150UL
Catalog # 50-172-6143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBC13 Fusion Protein Ag0292 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bendless protein; bendless-like ubiquitin conjugating enzyme; Bendless-like ubiquitin-conjugating enzyme; BLU UBC13 UbcH ben UBE2N Ubiquitin carrier protein N Ubiquitin protein ligase N; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; Ubc13; UbcH13; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2N; Ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); Ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Aliases: 1500026J17Rik; AL022654; BB101821; BLU; HEL-S-71; UBC13; UbcH-ben; UbcH13; UBCHBEN; UBC13; UBE2N
UniProt ID: (Human) P61088, (Rat) Q9EQX9, (Mouse) P61089
Entrez Gene ID: (Human) 7334, (Rat) 116725, (Mouse) 93765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.