Novus Biologicals
Manufacturer Code:NBP298559F
Catalog # NB104510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human UBAP1 fragment. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC119669 MGC8710 NAG20 Nasopharyngeal carcinoma-associated gene 20 protein UAP UBAP UBAP-1 ubiquitin associated protein ubiquitin associated protein 1 ubiquitin-associated protein 1; Nasopharyngeal carcinoma-associated gene 20 protein; UBAP-1; Ubiquitin-associated protein 1
Gene Aliases: NAG20; UAP; UBAP; UBAP-1; UBAP1
UniProt ID: (Human) Q9NZ09
Entrez Gene ID: (Human) 51271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.