Proteintech
Manufacturer Code:123851AP150UL
Catalog # 50-172-7945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBAP1 Fusion Protein Ag3055 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NAG20 UAP UBAP UBAP 1 UBAP1 ubiquitin associated protein 1; Nasopharyngeal carcinoma-associated gene 20 protein; UBAP; UBAP-1; Ubiquitin-associated protein 1
Gene Aliases: 2700092A01Rik; NAG20; UAP; UBAP; UBAP-1; UBAP1
UniProt ID: (Human) Q9NZ09, (Rat) Q5XIS7, (Mouse) Q8BH48
Entrez Gene ID: (Human) 51271, (Rat) 362502, (Mouse) 67123
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.