Invitrogen
Manufacturer Code:PA525216
Catalog # PIPA525216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 63-91 amino acids from the N-terminal region of human UBAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHGDHL1 RP11-178C10.1 UBA domain-containing protein 2 phosphoglycerate dehydrogenase like 1 phosphoglycerate dehydrogenase-like protein 1 ubiquitin-associated domain-containing protein 2; phosphoglycerate dehydrogenase like 1; Phosphoglycerate dehydrogenase-like protein 1; RP11-178C10.1; UBA domain-containing protein 2; Ubiquitin-associated domain-containing protein 2
Gene Aliases: 1190008A14Rik; AW554959; PHGDHL1; PSEC0110; UBAC2
UniProt ID: (Human) Q8NBM4, (Mouse) Q8R1K1
Entrez Gene ID: (Human) 337867, (Mouse) 68889
Molecular Function:
aspartic protease
hydrolase
protease
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.