Novus Biologicals
Manufacturer Code:NBP288529
Catalog # NB036151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human UBAC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase subunit KPC2; E3 ubiquitin-protein ligase subunit KPC2 GBDR1UBA domain-containing protein 1 Glialblastoma cell differentiation-related protein 1 Kip1 ubiquitination-promoting complex protein 2 KPC2 putative glialblastoma cell differentiation-related protein RP11-432J22.3 UBA domain containing 1 UBADC1 ubiquitin associated domain containing 1 ubiquitin-associated domain-containing protein 1; Glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; putative glialblastoma cell differentiation-related protein; testicular secretory protein Li 64; UBA domain-containing protein 1; ubiquitin associated domain containing 1; Ubiquitin-associated domain-containing protein 1
Gene Aliases: GBDR1; KPC2; UBAC1; UBADC1
UniProt ID: (Human) Q9BSL1
Entrez Gene ID: (Human) 10422
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.