Proteintech
Manufacturer Code:132111AP150UL
Catalog # 50-172-8575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UBA6 Fusion Protein Ag4000 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E1 L2 Monocyte protein 4 MOP 4 MOP4 UBA6 UBE1L2 Ubiquitin activating enzyme 6; E1-L2; Monocyte protein 4; MOP-4; UBA6, ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme 6; ubiquitin-activating enzyme E1-like 2; Ubiquitin-activating enzyme E1-like protein 2; ubiquitin-like modifier activating enzyme 6; Ubiquitin-like modifier-activating enzyme 6
Gene Aliases: 4930542H01; 5730469D23Rik; AU021846; AW124799; E1-L2; MOP-4; MOP4; RGD1308324; UBA6; UBE1L2
UniProt ID: (Human) A0AVT1, (Mouse) Q8C7R4
Entrez Gene ID: (Human) 55236, (Rat) 305268, (Mouse) 231380
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.