Invitrogen
Manufacturer Code:MA526849
Catalog # PIMA526849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F2 |
Immunogen | Synthetic peptide corresponding to residues near C-terminus of UBA52 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L40; CEP52; CEP52 HUBCEP52 L40 RPL40 60S ribosomal protein L40 ubiquitin carboxyl extension protein 52 ubiquitin-52 amino acid fusion protein ubiquitin-60S ribosomal protein L40 ubiquitin-CEP52; Large ribosomal subunit protein eL40; Ubiquitin; Ubiquitin A-52 residue ribosomal protein fusion product 1; ubiquitin carboxyl extension protein 52; ubiquitin-52 amino acid fusion protein; Ubiquitin-60S ribosomal protein L40; ubiquitin-CEP52
Gene Aliases: CEP52; HUBCEP52; L40; RPL40; UBA52; UBCEP2
UniProt ID: (Human) P62987
Entrez Gene ID: (Human) 7311
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.