OriGene
Manufacturer Code:TA811882
Catalog # 50-168-4314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-463 of human UBA3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NAE2 UBE1C hUBA3; NEDD8-activating enzyme E1 catalytic subunit; NEDD8-activating enzyme E1 subunit 2; NEDD8-activating enzyme E1C; Nedd8-activating enzyme hUba3; UBA3, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 3; Ubiquitin-activating enzyme E1C; ubiquitin-activating enzyme E1C (homologous to yeast UBA3); ubiquitin-activating enzyme E1C (UBA3 homolog, yeast); ubiquitin-like modifier activating enzyme 3; Ubiquitin-like modifier-activating enzyme 3
Gene Aliases: A830034N06Rik; AI256736; AI848246; AW546539; hUBA3; NAE2; UBA3; UBE1C
UniProt ID: (Human) Q8TBC4, (Mouse) Q8C878, (Rat) Q99MI7
Entrez Gene ID: (Human) 9039, (Dog) 476560, (Mouse) 22200, (Rat) 117553
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.