Invitrogen
Manufacturer Code:MA536102
Catalog # PIMA536102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B6-6-5 |
Immunogen | Recombinant protein within human UBA3 aa 200-463. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A830034N06Rik AI256736 AI848246 AW546539 DKFZp566J164 hUBA3 mgC22384 NAE2 NEDD8-activating enzyme E1 catalytic subunit NEDD8-activating enzyme E1 subunit 2 NEDD8-activating enzyme E1C Nedd8-activating enzyme hUba3 Uba3 UBA3 ubiquitin-activating enzyme E1 homolog Ube1c ubiquitin like modifier activating enzyme 3 ubiquitin-activating enzyme 3 Ubiquitin-activating enzyme E1C ubiquitin-activating enzyme E1C (homologous to yeast UBA3) ubiquitin-activating enzyme E1C (UBA3 homolog yeast) ubiquitin-like modifier activating enzyme 3 ubiquitin-like modifier-activating enzyme 3; NEDD8-activating enzyme E1 catalytic subunit; NEDD8-activating enzyme E1 subunit 2; NEDD8-activating enzyme E1C; Nedd8-activating enzyme hUba3; UBA3, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 3; Ubiquitin-activating enzyme E1C; ubiquitin-activating enzyme E1C (homologous to yeast UBA3); ubiquitin-activating enzyme E1C (UBA3 homolog, yeast); ubiquitin-like modifier activating enzyme 3; Ubiquitin-like modifier-activating enzyme 3
Gene Aliases: hUBA3; NAE2; UBA3; UBE1C
UniProt ID: (Human) Q8TBC4, (Rat) Q99MI7
Entrez Gene ID: (Human) 9039, (Rat) 117553
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.