Invitrogen
Manufacturer Code:MA525437
Catalog # PIMA525437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A3 |
Immunogen | Full length human recombit protein of UBA2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anthracycline-associated resistance ARX; HRIHFB2115 ARX SAE2 SUMO-1 activating enzyme subunit 2 SUMO-activating enzyme subunit 2 SUMO1 activating enzyme subunit 2 UBA2 ubiquitin-activating enzyme E1 homolog anthracycline-associated resistance ARX ubiquitin-like 1-activating enzyme E1B; SUMO-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; SUMO1 activating enzyme subunit 2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin-like 1 (sentrin) activating enzyme E1B; Ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; Ubiquitin-like modifier-activating enzyme 2
Gene Aliases: ARX; EGK_10449; HRIHFB2115; SAE2; UBA2; UBLE1B
UniProt ID: (Human) Q9UBT2
Entrez Gene ID: (Rhesus monkey) 705808, (Dog) 476490, (Human) 10054, (Rat) 308508
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.