Invitrogen
Manufacturer Code:PA575187
Catalog # PIPA575187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 598-640 of Human SAE2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anthracycline-associated resistance ARX; SUMO-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; SUMO-activating enzyme subunit 2 Anthracycline-associated resistance ARX Ubiquitin-like 1-activating enzyme E1B Ubiquitin-like modifier-activating enzyme 2 UBA2 SAE2 UBLE1B HRIHFB2115; SUMO1 activating enzyme subunit 2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin-like 1 (sentrin) activating enzyme E1B; ubiquitin-like 1 (sentrin) activating enzyme subunit 2; Ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; Ubiquitin-like modifier-activating enzyme 2
Gene Aliases: AA986091; ARX; HRIHFB2115; SAE2; UBA1; UBA2; Ubl1a2; UBLE1B
UniProt ID: (Human) Q9UBT2, (Mouse) Q9Z1F9
Entrez Gene ID: (Human) 10054, (Rat) 308508, (Mouse) 50995
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.