Invitrogen
Manufacturer Code:PA5104470
Catalog # PIPA5104470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Uba2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA986091 Anthracycline-associated resistance ARX ARX FLJ13058 HRIHFB2115 Sae2 SUMO1 activating enzyme subunit 2 Sumo-1 activating enzyme subunit 2 SUMO-activating enzyme subunit 2 UBA1 Uba2 UBA2 ubiquitin-activating enzyme E1 homolog ubiquitin like modifier activating enzyme 2 ubiquitin-like 1 (sentrin) activating enzyme E1B ubiquitin-like 1 (sentrin) activating enzyme subunit 2 ubiquitin-like 1-activating enzyme E1B ubiquitin-like modifier activating enzyme 2 ubiquitin-like modifier-activating enzyme 2 Ubl1a2 Uble1b; Anthracycline-associated resistance ARX; SUMO-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; SUMO1 activating enzyme subunit 2; UBA2, ubiquitin-activating enzyme E1 homolog; Ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; Ubiquitin-like modifier-activating enzyme 2
Gene Aliases: ARX; HRIHFB2115; SAE2; UBA2; UBLE1B
UniProt ID: (Human) Q9UBT2
Entrez Gene ID: (Human) 10054
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.