Invitrogen
Manufacturer Code:PA566172
Catalog # PIPA566172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human UAP56 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 56 kDa U2AF65-associated protein; ATP-dependent RNA helicase p47; BAT1 D6S81E UAP56 56 kDa U2AF65-associated protein ATP-dependent RNA helicase p47 HLA-B-associated transcript 1 protein nuclear RNA helicase (DEAD family) spliceosome RNA helicase BAT1 spliceosome RNA helicase DDX39B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B; DEAD box protein UAP56; HLA-B-associated transcript 1 protein; nuclear RNA helicase (DEAD family); spliceosome RNA helicase BAT1; Spliceosome RNA helicase DDX39B
Gene Aliases: BAT1; D6S81E; DDX39B; UAP56
UniProt ID: (Human) Q13838
Entrez Gene ID: (Human) 7919
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.