Invitrogen
Manufacturer Code:PA569578
Catalog # PIPA569578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse UAP1L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: UAP1; UDP-N-acetylhexosamine pyrophosphorylase-like protein 1; UDP-N-acteylglucosamine pyrophosphorylase 1, like 1; UDP-N-acteylglucosamine pyrophosphorylase 1-like 1
Gene Aliases: 5730445F03Rik; UAP1L1; zgc:56509
UniProt ID: (Human) Q3KQV9, (Mouse) Q3TW96, (Zebrafish) Q7ZWD4
Entrez Gene ID: (Human) 91373, (Horse) 100067752, (Mouse) 227620, (Rat) 296560, (Zebrafish) 393264
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.