Invitrogen
Manufacturer Code:MA538172
Catalog # PIMA538172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1769 |
Immunogen | A synthesized peptide derived from human UAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AA420407 AA437972 AGX AGX1 AGX-1 AGX2 AGX-2 Antigen X ESTM38 RGD1561967 SPAG2 sperm associated antigen 2 sperm-associated antigen 2 testis tissue sperm-binding protein Li 37a UAP1 UDP-N-acetylgalactosamine pyrophosphorylase UDP-N-acetylglucosamine diphosphorylase 1 UDP-N-acetylglucosamine pyrophosphorylase UDP-N-acetylglucosamine pyrophosphorylase 1 UDP-N-acetylhexosamine pyrophosphorylase UDP-N-acetylhexosamine pyrophosphorylase 1 UDP-N-acteylglucosamine pyrophosphorylase 1; AGX; AGX-1; AGX-2; Antigen X; sperm associated antigen 2; Sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylglucosamine pyrophosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1
Gene Aliases: AA420407; AA437972; AGX; AGX-1; AGX-2; AGX1; AGX2; ESTM38; RGD1561967; SPAG2; UAP1
UniProt ID: (Human) Q16222, (Mouse) Q91YN5
Entrez Gene ID: (Human) 6675, (Mouse) 107652, (Rat) 498272
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.