Invitrogen
Manufacturer Code:PA531187
Catalog # PIPA531187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 232 of Human UAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGX; AGX AGX1 AGX2 SPAG2 UDP-N-acetylhexosamine pyrophosphorylase antigen X sperm associated antigen 2 sperm-associated antigen 2; AGX-1; AGX-2; Antigen X; sperm associated antigen 2; Sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1
Gene Aliases: AGX; AGX1; AGX2; SPAG2; UAP1
UniProt ID: (Human) Q16222
Entrez Gene ID: (Human) 6675
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.